GC Analysis of Phenylethanol Enantiomers on β-DEX™ 120
Conditions
column | beta-DEX 120, 30 m x 0.25 mm I.D., 0.25 µm (24304) |
oven | 120 °C |
carrier gas | helium, 20 cm/sec |
sample | 0.5 mg/mL each analyte in methylene chloride |
injection | 1 µL, 100:1 split |
inj. temp. | 200 °C |
detector | FID, 300 °C |
Description
Categories | Analytical Chromatography, GC Applications, Alcohols, Enantiomers |
Featured Industry | Chemicals and Industrial Polymers Flavors and Fragrances |
Legal Information | DEX is a trademark of Sigma-Aldrich Co. LLC |
Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
suitability | application for GC |