Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
OSHA Method 52: GC Analysis of Formaldehyde on SUPELCOWAX® 10 after Collection/Desorption using ORBO™-24
OSHA Method 52: GC Analysis of Formaldehyde on SUPELCOWAX® 10 after Collection/Desorption using ORBO™-24
Conditions
sample/matrix HMP-coated Supelpak 20 spiked with formaldehyde (10 mug)
adsorbent tube ORBO-24 (20231)
column SUPELCOWAX 10, 30m x 0.53 mm I.D., 0.5 µm (25325)
oven 50 °C (2 min) to 200 °C at 10 °C/min
carrier gas helium, 7 mL/min
injection 1 µLdirect, 220 °C
detector NPD, 220 °C
Description
Categories Analytical Chromatography, GC Applications, Air Monitoring Applications, Aldehydes & Ketones, Volatiles, Aldehydes
Featured Industry Industrial Hygiene
General description This is an application, not a product for sale.
See "Related Products" to find associated products for sale.
Or, for more information, email: Supelco Technical Service
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC, ORBO is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • 2-(Hydroxymethyl)piperidine (HMP)
  • Formaldehyde derivative