Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Desalting of Immunoglobulins (IgG, Human) on HiTrap® Desalting Cartridge
Desalting of Immunoglobulins (IgG, Human) on HiTrap® Desalting Cartridge
Conditions
column HiTrap Desalting Cartridge, 5 mL (54822)
mobile phase 0.2 M sodium carbonate buffer, pH 8.5
injection 0.5 mL unbound human IgG in 0.2 M sodium carbonate buffer after coupling to a HiTrap NHS 1 mL column. Fraction volume 0.5 mL
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information HiTrap is a registered trademark of GE Healthcare