Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis Carbohydrates on Proteomix® SAX-NP5
HPLC Analysis Carbohydrates on Proteomix® SAX-NP5
Conditions
column Proteomix SAX-NP5, 15 cm x 4.6 mm I.D., 5 µm particles (Z777228)
mobile phase [A] 0.05% (25% ammonium hydroxide) in acetonitrile; [B] 0.05% (25% ammonium hydroxide) in water
gradient 15 to 80% B in 20 min
flow rate 0.7 mL/min
temp. 35 °C
sample Corn starch hydrolysate; glucose oligomer (G1 to G6)
detector ELSD
Description
Categories Analytical Chromatography, Carbohydrates
Featured Industry Food and Beverages
Vitamins, Nutraceuticals, and Natural Products
Legal Information Proteomix is a registered trademark of Sepax Technologies