Attention:
We are currently upgrading the certain systems that support this website.  You may experience some functionality loss, though we expect the site to be fully functional by 9:00PM CT (US), February 24th.
We apologize for any inconvenience and appreciate your patience during this improvement process.Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
HPLC Analysis of 16 Triazine and Phenyl Urea Compounds on Ascentis® Express RP-Amide, Modified Gradient
Conditions
column Ascentis Express RP-Amide, 15 cm x 3 mm I.D., 2.7 µm particles (53919-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile
gradient 10 to 25% B in 30 min; 25 to 60% B in 15 min; held at 60% B for 5 min
flow rate 0.4 mL/min
sample 20 mg/L each (except terbuthylazin desethyl and terbuthylazine 60 mg/L) in 60:40, water: acetonitrile
injection 5 µL
detector UV, 240 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express RP-Amide for the efficient separation of 16 triazine and phenyl urea compounds. Lower column
temperature can improve resolution of the critical pair.
Categories Analytical Chromatography, Pesticides & Herbicides
Featured Industry Agriculture
Chemicals and Industrial Polymers