Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Albuterol Enantiomers on Astec® CHIROBIOTIC® T2
HPLC Analysis of Albuterol Enantiomers on Astec® CHIROBIOTIC® T2
Conditions
column CHIROBIOTIC T2, 25 cm x 4.6 mm I.D., 5 µm particles (16024AST)
column temp. 25 °C
mobile phase 0.1 wt% ammonium trifluoroacetic acid in methanol
flow rate 1 mL/min
sample 1 mg/mL in methanol
injection 1 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Bronchodilators, Pharmaceutical (small molecule), All Pharmaceutical Applications