Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Anticonvulsant Drugs on Discovery® RP-AmideC16
HPLC Analysis of Anticonvulsant Drugs on Discovery® RP-AmideC16
Conditions
column Discovery C18, 15 cm x 4.6 mm I.D., 5 µm particles (504955)
column temp. 40 °C
mobile phase [A] acetonitrile; [B] 25 mM potassium phosphate, pH 7.0 (40:60, A:B)
flow rate 1 mL/min
injection 10 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Clinical, Pharmaceutical Analysis, antiepileptic/antiseizure, anticonvulsants, Anticonvulsant/antiepiletic/antiseizure
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC