Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Antipsychotic Drugs on Ascentis® Express RP-Amide
HPLC Analysis of Antipsychotic Drugs on Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 µm particles (53913-U)
column temp. 35 °C
mobile phase [A] 5 mM ammonium formate (in water); [B] 5 mM ammonium formate (in 95:5 methanol:water)
flow rate 3 mL/min
sample 200 µg/L in water
injection 1 µL
detector ESI(+), TIC 100 - 1000 m/z
Description
Categories Analytical Chromatography, Clinical, antipsychotics, Antipsychotics
Featured Industry Clinical
Pharmaceutical (small molecule)