Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Bile Acids on Ascentis® Express C18
HPLC Analysis of Bile Acids on Ascentis® Express C18
Conditions
column Ascentis Express C18, 5 cm x 4.6 mm I.D., 2.7 µm particles (53826-U)
column temp. 35 °C
mobile phase [A] 10 mM ammonium acetate, pH 4 (adjusted with acetic acid); [B] acetonitrile; (80:20, A:B)
gradient 20 to 100% B in 10 min
flow rate 1.0 mL/min
sample 10 µg/mL
injection 5 µL
detector ESI(-) in SIR mode
Description
Categories Analytical Chromatography, Clinical, Organic Acids, organic acids