Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Bisphenol A, Dimethylbisphenol A and Nonylphenol on Ascentis® Express RP-Amide (UV Detection)
HPLC Analysis of Bisphenol A, Dimethylbisphenol A and Nonylphenol on Ascentis® Express RP-Amide (UV Detection)
Conditions
column Ascentis Express RP-Amide, 5 cm x 4.6 mm I.D., 2.7 µm particles (53922-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile
gradient 50 to 100% B in 10 min
flow rate 1.0 mL/min
sample 50 mg/L of each in 50:50, water: acetonitrile
injection 10 µL
detector UV, 280 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis Express C18 for the efficient separation of milk thistle related compounds
Categories Analytical Chromatography, Food & Beverage Analysis, Environmental Analysis, Phenols, Beverage Testing, Organics, Milk, Drinking Water and Bottled Water, Chromatograms, HPLC Chromatograms
Featured Industry Environmental