Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Bronchodilators Theophylline and Other Caffeine Metabolites in Serum on Discovery® RP-AmideC16 after SPE using Discovery® DSC-18
HPLC Analysis of Bronchodilators Theophylline and Other Caffeine Metabolites in Serum on Discovery® RP-AmideC16 after SPE using Discovery® DSC-18
Conditions
sample preparation 1 mL porcine serum spiked with 0.1 µg/mL, 0.50 µg/mL, or 1.0 µg/mL each analyte
sample preparation 2 mL 5% methanol in water; dry tube 10 min with nitrogen stream
sample preparation 1 mL methanol; evaporate to dryness with nitrogen stream at room temperature; reconstitute in 200 µL mobile phase containing 0.2 µg/mL or 7-methylxanthine (IS)
sample preparation SPE (Solid Phase Extraction)
SPE tube/cartridge Discovery® DSC-18, 500 mg/3 mL (52603-U)
condition 2 mL methanol, then 2 mL water
column Discovery RP-AmideC16, 15 cm x 4.6 mm, 5 µm particles, preceded by a 2 cm RP-AmideC16 guard column and 0.5 µm frit filter (505013)
column temp. 30 °C
mobile phase [A] Methanol; [B] 1% acetic acid (17:38, A:B)
flow rate 1 mL/min
injection 20 µLreconstituted porcine serum extract
detector UV, 272 nm
Description
Categories Analytical Chromatography, SPE Applications, Bronchodilators, Pharmaceutical & Clinical applications
Featured Industry Clinical
Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for SPE, application for HPLC