Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Chloropheniramine Maleate & Related Impurities on Chromolith® High Resolution RP18e
HPLC Analysis of Chloropheniramine Maleate & Related Impurities on Chromolith® High Resolution RP18e
Conditions
column Chromolith High Resolution RP18 endcapped, 100 x 4.6 mm (1.52022.0001)
mobile phase [A] 8.57 gm ammonium dihurogen phosphate in 1 lit water. pH adjusted to 3.0 with
OPA; [B] acetonitrile: (80:20, A:B)
flow rate 1.2 mL/min
column temp. 25 °C
detector 225 nm cell: 10 μL
injection 5 μL
sample 0.100 gm substance in 100 ml mobile phase.
Description
Categories Analytical/Chromatography, Applications, HPLC Applications, Pharmaceuticals
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany