Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Diclofenac and Salicylic Acid on Ascentis® RP-Amide
HPLC Analysis of Diclofenac and Salicylic Acid on Ascentis® RP-Amide
Conditions
column Ascentis RP-Amide, 15 cm x 4.6 mm I.D., 5 µm particles (565324-U)
column temp. 35 °C
mobile phase [A] 20 mM phosphoric acid (pH 2.0 unadjusted); [B] acetonitrile; (50:50, A:B)
flow rate 1.0 mL/min
sample as indicated in 20 mM phosphoric acid (pH 2.0)
injection 10 µL
detector UV, 220 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Anti-inflammatories