Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Diquat on SeQuant ZIC-HILIC. Effect of %Organic in the Mobile Phase on Retention
HPLC Analysis of Diquat on SeQuant ZIC-HILIC. Effect of %Organic in the Mobile Phase on Retention
Conditions
column SeQuant ZIC-HILIC, PEEK, 150 x 4.6 mm, 5 µm, 200 Ã… (1.50455.0001)
column temp. 40 °C
mobile phase [A] acetonitrile; [B] 250 mM ammonium acetate, pH 3.7 (see Figure for ratio)
flow rate 1.0 mL/min
sample 50 ppm diquat in acetonitrile:water (1:1)
injection 5 µL
detector 313 nm
Description
Featured Industry Agriculture
General Analytical