Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Fluoxetine Enantiomers on Narrowbore Astec® CHIROBIOTIC® V2
HPLC Analysis of Fluoxetine Enantiomers on Narrowbore Astec® CHIROBIOTIC® V2
Conditions
column CHIROBIOTIC V2, 10 cm x 2.1 mm I.D., 5 µm particles (15018AST)
column temp. 25 °C
mobile phase 13 mM ammonium acetate in methanol
flow rate 0.2 mL/min
sample 1 mg/mL in methanol
injection 5 µL
detector UV, 230 nm
Description
Categories Analytical Chromatography, Chiral Applications, Antidepressants, Pharmaceutical (small molecule), All Pharmaceutical Applications