Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Ginsenosides on Ascentis® Express C18
HPLC Analysis of Ginsenosides on Ascentis® Express C18
Conditions
column Ascentis Express C18, 15 cm x 4.6 mm I.D., 2.7 µm particles (53829-U)
column temp. 35 °C
mobile phase [A] water; [B] acetonitrile
gradient 25% B for 1.5 min; 25 to 85% B in 12 min; hold at 85% B for 1 min
flow rate 1 mL/min
sample 50 µg/mL in 82:18, water:methanol
injection 5 µL
detector UV, 254 nm
Description
Categories Analytical Chromatography, Glycosides
Featured Industry Vitamins, Nutraceuticals, and Natural Products