Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Glutathione S-Transferase on TSKgel® G3000SWXL
HPLC Analysis of Glutathione S-Transferase on TSKgel® G3000SWXL
Conditions
column TSKgel G3000SWXL, 30 cm x 7.8 mm I.D., 5 µm particles (808541)
column temp. 25 °C
mobile phase 0.3M sodium chloride/0.05M sodium dihydrogen phosphate, pH 7.0
flow rate 1 mL/min
injection 100µL crude glutathione S-transferase preparation from guinea pig liver (0.7mg)
detector UV, 220nm and enzyme assay
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation