HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L and CLC-D
Conditions
column | Astec CLC-L and CLC-D, 15 cm x 4.6 mm I.D., 5 µm particles (53023AST, 53123AST) |
column temp. | 25 °C |
mobile phase | 5 mM copper sulfate |
flow rate | 1 mL/min |
injection | 10 µL |
detector | UV, 254 nm |
Description
Categories | Analytical Chromatography, Chiral Applications, Chemicals and Industrial Polymers, Organic Acids |
Featured Industry | Chemicals and Industrial Polymers Vitamins, Nutraceuticals, and Natural Products |
Legal Information | Astec is a registered trademark of Sigma-Aldrich Co. LLC |
suitability | application for HPLC |
Materials