Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Lipoxidase on TSKgel® Phenyl-5PW
HPLC Analysis of Lipoxidase on TSKgel® Phenyl-5PW
Conditions
column TSKgel Phenyl-5PW, 7.5 cm x 7.5 mm I.D., 10 µm particles (807573)
mobile phase [A] 0.1M phosphate buffer, pH 7.0 + 1.5M ammonium sulfate; [B] 0.1M phosphate buffer, pH 7.0;
gradient linear, 0% B to 100% B over 60 min
flow rate 0.5 mL/min
detector UV, 280 nm
Description
Categories Analytical Chromatography, Proteins
Featured Industry Life Science and Biopharma
Legal Information TSKgel is a registered trademark of Tosoh Corporation