Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of α-Methyl-α-Phenylsuccinimide Enantiomers on Astec® CHIROBIOTIC® T
HPLC Analysis of α-Methyl-α-Phenylsuccinimide Enantiomers on Astec® CHIROBIOTIC® T
Conditions
column CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 µm particles (12024AST)
column temp. 23 °C
mobile phase [A] ethanol; [B] hexane; (20:80, A:B)
flow rate 3 mL/min
sample 5 mg/mL in methanol
injection 5 µL
detector UV, 210 nm
Description
Categories Analytical Chromatography, Chiral Applications, Organic Bases, Chemicals and Industrial Polymers, Amines