HPLC Analysis of Monosaccharides - Ribose, Xylitol, Mannose on Chromolith(R) NH2 100 - 4.6mm
HPLC Analysis of Monosaccharides - Ribose, Xylitol, Mannose on Chromolith® NH2 100 - 4.6mm
Conditions
column Chromolith® NH2, 100 x 4.6 mm (1.52028.0001)
column temp. 23 °C
mobile phase [A] acetonitrile; [B] water; (85:15, A:B)
flow rate 3.0 mL/min
pressure 682 psi (47 bar)
sample Ribose 5.9 mg/mL, Xylitol 8.1mg/mL, Mannose 7.6mg/mL in mobile phase
injection 5 μL
detector 190 nm
Description
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany