HPLC Analysis of Rapamycin and 32-Desmethoxyrapamycin on Discovery® HS C18
Conditions
column | Discovery HS C18, 15 cm x 4.6 mm ID, 3mum particles (568522-U) |
column temp. | 60 °C |
mobile phase | [A] 10 mM ammonium acetate, pH unadjusted: [B] acetonitrile (25:75, v/v, A:B) |
flow rate | 1.5 mL/min |
injection | 10 µg/mL |
detector | UV, 268 nm |
Description
Categories | Analytical Chromatography, Clinical, immunosuppressants, Immunosuppressants |
Featured Industry | Clinical |
Legal Information | Discovery is a registered trademark of Sigma-Aldrich Co. LLC |
suitability | application for HPLC |
Materials