Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Sulfonamide Positional Isomers on Discovery® Zr-Carbon C18
HPLC Analysis of Sulfonamide Positional Isomers on Discovery® Zr-Carbon C18
Conditions
column Discovery Zr-Carbon C18, 15 cm x 4.6 mm I.D., 3 µm particles (65706-U)
column temp. 80 °C
mobile phase [A] 10mM diethylamine, pH 10.8; [B] acetonitrile;
flow rate 1.5 mL/min
injection 5 µL
detector UV, 240 nm
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics
Featured Industry Chemicals and Industrial Polymers
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC