HPLC Analysis of Tetraconazole Enantiomers on Astec(R) Cellulose DMP in Normal Phase Mode
HPLC Analysis of Tetraconazole Enantiomers on Astec(R) Cellulose DMP in Normal Phase Mode
Description
Analysis Note The fungicide tetraconazole is a chiral molecule, comprising two enantiomers. Enantioselective influences of chiral pesticides and their metabolites on ecosystems is an area of great importance. The possibility that only one enantiomer has the wanted properties and/or the other form an unwanted effect may exist. An analytical separation of the enantiomers is required to perform studies in these areas. The ASTEC® Cellulose DMP column resolved the enantiomers of tetraconazole under normal phase conditions.
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC