Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Triazole Antifungal Agent Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
HPLC Analysis of Triazole Antifungal Agent Enantiomers on Astec® CYCLOBOND® I 2000 HP-RSP
Conditions
column Astec CYCLOBOND I 2000 HP-RSP, 25 cm x 4.6 mm I.D., 5 µm particles (24024AST)
mobile phase [A] 0.1% formic acid; [B] acetonitrile; (70:30, A:B)
flow rate 1 mL/min
sample 0.1 mg/mL in methanolpeaks 1&2: enilconazolepeaks 2&3: eberconazolepeaks 4&5: miconazolepeaks 6&7: ketoconazole
injection 1 µL
detector UV, 230 nm
Description
Analysis Note Note: Peak 2 of enilconzaole coelutes with peak 1 of eberconazole under these conditions.
Categories Analytical Chromatography, Chiral Applications, Antibiotics, Antimicrobials, Pharmaceutical (small molecule), Antifungals, All Pharmaceutical Applications