Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Tricyclic Antidepressants on Ascentis® ES Cyano
HPLC Analysis of Tricyclic Antidepressants on Ascentis® ES Cyano
Conditions
column Ascentis ES Cyano, 15 cm x 4.6 mm I.D., 5 µm particles (577306-U)
column temp. 25 °C
mobile phase [A] 10 mM potassium phosphate dibasic, pH 7 with phosphoric acid; [B] acetonitrile; [C] methanol; (25:60:15, A:B:C)
flow rate 2 mL/min
sample 100 µg/mL in water: methanol (70:30)
injection 5 µL
detector UV, 215 nm
Description
Analysis Note This application demonstrates the suitability of Ascentis ES Cyano for the efficient separation of tricyclic antidepressants.
Categories Analytical Chromatography, Antidepressants