Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Veterinary Antibiotics on Discovery® HS C18
HPLC Analysis of Veterinary Antibiotics on Discovery® HS C18
Conditions
column Discovery HS C18, 5 cm x 2.1 mm I.D., 3 µm particles (569253-U)
column temp. 22 °C
mobile phase [A] (50 mM acetic acid/ammonium hydroxide, pH 6.0]: [B] methanol; (20:80, A:B):
flow rate 0.2 mL/min
sample as indicated in 50:50 water:methanol
injection 2 µL
detector ESI (+), full scan
Description
Categories Analytical Chromatography, Pharmaceutical Analysis, Antibiotics, Antibiotics, Antimicrobials
Featured Industry Pharmaceutical (small molecule)
Legal Information Discovery is a registered trademark of Sigma-Aldrich Co. LLC