Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
HPLC Analysis of Vitamins B1, B6 and B12 using Ascentis® Express RP-Amide
HPLC Analysis of Vitamins B1, B6 and B12 using Ascentis® Express RP-Amide
Conditions
column Ascentis Express RP-Amide, 10 cm x 4.6 mm I.D., 5 µm particles (50773-U)
column temp. 30 °C
mobile phase [A] 0.02 M Ammonium Formate, pH = 3.0; [B] acetonitrile
gradient 0-50% B in 1 min. Hold at 50% B until 1.5 min
flow rate 1.5 mL/min
pressure 1639 psi (113 bar)
sample Dissolved in 50/50 water/ACN
injection 1 µL
detector UV, 276 nm
Description
Categories Analytical Chromatography, Vitamins and Natural products
Featured Industry Vitamins, Nutraceuticals, and Natural Products