Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
LC/MS Analysis of Green Tea Components on Chromolith® FastGradient RP-18 endcapped
LC/MS Analysis of Green Tea Components on Chromolith® FastGradient RP-18 endcapped
Conditions
column Chromolith FastGradient RP-18, 5 cm x 2.0 mm I.D., 2 µm particles (1.52007.0001)
column temp. 25 °C
mobile phase [A] 0.1% formic acid in acetonitrile; [B] 0.1% formic acid in water
gradient 100% B for 0.6 min, to 72.5% B in 7.4 min, held at 72.5% for 1 min
flow rate 0.4 µL/min
pressure 605 psi (42 bar)
injection 1 µL
detector ESI-MS(+) (m/z range 150-950), BPC and overlay of EICs)
Description
Categories Analytical Chromatography, Natural products
Featured Industry Food and Beverages
Legal Information Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany
Other Notes Sample Prep: 1.75 g of green tea was extracted for three minutes using 100 mL of boiling water. Prior to injection both samples were centrifuged at 4500 rpm for 10 min and cleaned using a 0.45 um syringe filter.
suitability application for LC-MS