Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
UHPLC Analysis of IgM from Human Serum on BIOshell™ A400 Protein C4
UHPLC Analysis of IgM from Human Serum on BIOshell™ A400 Protein C4
Conditions
column BIOshell A400 Protein C4; 10 cm x 2.1 mm I.D., 3.4 µm particles (66825-U)
column temp. 75 °C
mobile phase [A] 0.1% TFA in water; [B] 0.1% TFA in acetonitrile
gradient 30 to 60% B in 10 min; 60 to 100% B in 10 min
flow rate 0.2 mL/min
pressure 520 psi (36 bar)
sample IgM, 1 g/L, 0.05% TFA in water
injection 1 µL
detector UV, 215 nm
Description
Analysis Note Immunoglobulin M (IgM) antibodies are pentameric complexes, produced by B cells, that are initially deployed by the immune system as a response to antigens. IgM has a molecular weight of around 970 kDa. Chromatographic analysis of this antibody was performed using a BIOshell A400 Protein C4 column.
Categories Analytical Chromatography, Peptides, Antibodies, Proteins
Featured Industry Life Science and Biopharma
Legal Information BIOshell is a trademark of Sigma-Aldrich Co. LLC