Analytical Method: Iron in filter media
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114549  Spectroquant® Iron Cell Test
Cat.No. 114761  Spectroquant® Iron Test
Cat.No. 100796  Spectroquant® Iron Test
Cat.No. 100244  Citric acid for analysis
Cat.No. 116754  Water for analysis
Folded filter
Citric acid 5 % aqueous solution:
Dissolve 5 g Citric acid for analysis with 100 ml water for analysis.
Sample preparation:
Stir mechanically 5 g of the material with 100 ml of 5 % citric acid solution (30 min). After filtration through a folded filter determine the iron content directly.
Determine with the above mentioned test kits.
Iron content in mg/kg Fe = analysis value in mg/l Fe x 20
Related Links