Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
Analytical Method: Nitrate in disposal drainage water
Instruments and Reagents:
Cat.No. 173016  Spectroquant® VIS Spectrophotometer Prove 100 or
Cat.No. 173017  Spectroquant® UV/VIS Spectrophotometer Prove 300 or
Cat.No. 173018  Spectroquant® UV/VIS Spectrophotometer Prove 600 or
Cat.No. 109748  Spectroquant® Photometer NOVA 30 or
Cat.No. 109751  Spectroquant® Photometer NOVA 60 or
Cat.No. 173632  Spectroquant® Colorimeter Move 100
Cat.No. 114946  Rectangular cells 10 mm and/or
Cat.No. 114947  Rectangular cells 20 mm and/or
Cat.No. 114944  Rectangular cells 50 mm
Cat.No. 114542  Spectroquant® Nitrate Cell Test
Cat.No. 114773  Spectroquant®  Nitrate Test
Cat.No. 116754  Water for analysis
Sample preparation - hot water extraction:
Use the original sample or the dilution (about 1:10) respectively. With undiluted samples their own color or discoloration because of a high part of organic material can be neutralized by determining a calibration value, made of the sample solution and reagent NO3-1 without addition of reagent NO3-2, against a blank reading. This value will then be substrated from the actual analysis value.
Analysis:
Determine with the above mentioned test kits.
Calculation:
Nitrate content in mg/l NO3 = analysis value in mg/l NO3 minus blank reading
or for 1:10 - dilution: (analysis value minus blank reading) x 10