TLC Analysis Of A Natural Product From Cosmetical Matrix on HPTLC Silica gel 60 NH2 F254s
TLC Analysis Of A Natural Product From Cosmetical Matrix on HPTLC Silica gel 60 NH2 F254s
Conditions
sample preparation Standard was diluted in 10 mL isopropanol,sample was directly applied
to the plate by CAMAG ATS4 6 mm bandwise.
plate 1.15161 HPTLC Silica gel 60 RP-18 F 254s MS-grade 20cm x 10 cm (1.151161.0001)
eluent [A] methanol; [B] acetonitrile; (9:1, A:B)
migration distance 6 cm
chamber normal chamber without chamber saturation
sample 0.150 μL
detector UV, 254 nm (Identification by coupling the TLC – MS Interface (CAMAG) with the Advion
expression)(CMS mass spectrometer -ESI mode.
Eluent: Acetonitrile / Water 95:5 + 0.1% TFA.)
Description
suitability application for TLC