TLC Analysis of Adenosine Phosphates on HPTLC NH2 F254s
TLC Analysis of Adenosine Phosphates on HPTLC NH2 F254s
Conditions
plate  1.13192 HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent [A] methanol; [B] water; (30:70, A:B) containing 0,5g NaCl
migration distance  5 cm
sample  0.2 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm.