TLC Analysis of Anabolic Compounds on HPTLC DIOL F254s (analyte set 1)
TLC Analysis of Anabolic Compounds on HPTLC DIOL F254s (analyte set 1)
Conditions
plate 1.05636 HPTLC pre-coated plate DIOL F 254s (1.05636.0001, 1.12668.0001)
eluent [A] di-iso-propyl ether; [B] acetic acid glacial; (100:1, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.3 μL
Description
suitability application for TLC