TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-2 F254s (analyte set 1)
TLC Analysis of Chlorophenols on HPTLC Silica Gel 60 RP-2 F254s (analyte set 1)
Conditions
plate: 1.05747 HPTLC pre-coated plate RP-2 F 254s (1.05747.0001)
eluent: [A] methanol; [B] water; (70:30, A:B)
migration distance: 5 cm
chamber: normal chamber without chamber saturation
sample: 0.3 μL
detector: in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC