TLC Analysis of Dinitrophenols on HPTLC pre-coated plate RP 18W F254s (analyte set 2)
TLC Analysis of Dinitrophenols on HPTLC pre-coated plate RP 18W F254s (analyte set 2)
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s (1.13724.0001)
eluent [A] acetone; [B] water; (30:70, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.1 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC