TLC Analysis of Expectorants and Related Products on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Expectorants and Related Products on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
instrument Camag TLC/HPTLC scanner
plate HPTLC pre-coated plate RP-18w F 254s
eluent   [A] acetonitrile; [B] water; (30:70, A:B) containing 0.1 M TEACI 
migration distance   7 cm
chamber  normal chamber without chamber saturation
sample application volume 0.3 µL (300 nL)
detection in situ evaluation with TLC/HPTLC scanner (Camag) UV 220 nm
Description
Featured Industry Pharmaceutical (Small Molecule)
Analysis Note In situ evaluation with TLC/HPTLC scanner
Application No. ASC120118C