Attention:
Certain features of sigmaaldrich.com will be down for maintenance Saturday afternoon/evening, February 24th starting at 3:00 pm CT until 9:00 pm CT.
Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
TLC Analysis of Fatty Acids as Isatinmethyl Esters on HPTLC Silica Gel 60 RP-18 F254s
TLC Analysis of Fatty Acids as Isatinmethyl Esters on HPTLC Silica Gel 60 RP-18 F254s
Conditions
detector in-situ evaluation with TLC/HPTLC scanner (Camag) UV 313 nm
plate HPTLC pre-coated plate RP-18 F 254s (1.15423.0001, 1.15389.0001)
chamber normal chamber without chamber saturation
eluent [A] methanol; [B] water; (88:12, A;B)
Description
Categories Analytical Chromatography, TLC Applications, fatty acids methyl esters
Featured Industry Food and Beverages
Life Science and Biopharma
General description TLC Plate Information: 1.13724.0001
suitability application for TLC