TLC Analysis of ingredients in Cardiac Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone
TLC Analysis of ingredients in Cardiac Agents on HPTLC Silica Gel 60 RP-18 WF 254s with concentrating zone
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s with concentrating zone (1.13124.0001)
eluent [A] methanol; [B] water; [C] acetonitrile; (50:50:30, A:B:C)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 5 μL
detector Dipped in MnCl2-sulfuric acid heated up to 110° C for 5 min (in situ evaluation with
TLC/HPTLC scanner (Camag) UV 248 nm. )
Description
Featured Industry Life Science and Biopharma
suitability application for TLC