TLC Analysis of Nitrophenols on HPTLC Pre-Coated Plate CN F254s
TLC Analysis of Nitrophenols on HPTLC Pre-Coated Plate CN F254s
Conditions
plate: HPTLC pre-coated plate CN F 254s (1.16464.0001)
eluent: [A] carbon tetrachloride; [B] methanol; (48:2, A:B) migration distance: 7 cm
chamber: normal chamber without chamber saturation
sample: 0.2 μL
detector: (in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm) suitability application for TLC Featured Industry Chemicals and Industrial Polymers