TLC Analysis of Nucleobases on HPTLC Silica Gel 60 RP-2 F254s (analyte set 1)
TLC Analysis of Nucleobases on HPTLC Silica Gel 60 RP-2 F254s (analyte set 1)
Conditions
plate 1.05747 HPTLC pre-coated plate RP-2 F 254s (1.05747.0001)
eluent Acetonitrile/water 60/40 (v/v)
migration distance 7 cm
chamber normal chamber without chamber saturation
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 264 nm
Description
suitability application for TLC
Featured Industry
Life Science and Biopharma