TLC Analysis of p-Hydroxybenzoic Acid and p-Hydroxybenzoic Acid Esters on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of p-Hydroxybenzoic Acid and p-Hydroxybenzoic Acid Esters on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
plate HPTLC pre-coated plate RP-18 wF 254s (1.13124.0001)
eluent [A] acetone; [B] water; (50:50, A:B)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.1 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 270 nm
Description
Featured Industry Chemicals and Industrial Polymers
suitability application for TLC