TLC Analysis of Plant-Protective Agents on HPTLC Silica Gel 60 F 254
TLC Analysis of Plant-Protective Agents on HPTLC Silica Gel 60 F 254
Conditions
plate 1.05642 HPTLC pre-coated plate silica gel 60 F 254 (1.05642.0001, 1.05548.0001)
eluent n-heptane
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.2 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
suitability application for TLC
Featured Industry Agriculture