TLC Analysis of Polycyclic Aromatic Hydrocarbons on HPTLC Silica Gel 60 RP-18 WF254s
TLC Analysis of Polycyclic Aromatic Hydrocarbons on HPTLC Silica Gel 60 RP-18 WF254s
Conditions
plate HPTLC pre-coated plate RP-18 F 254s with concentrating zone(1.15498.0001)
eluent [A] acetonitrile; [B] methylene chloride; [C] water; (90:10:10 A:B:C)
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 5 μL
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 386 nm
Description
Featured Industry Environmental
suitability application for TLC