TLC Analysis of Purine Derivatives on HPTLC NH2 F254s (analyte set 1)
TLC Analysis of Purine Derivatives on HPTLC NH2 F254s (analyte set 1)
Conditions
plate  HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent [A] ethanol; [B] water; (80:20, A:B) saturated with NaCl
migration distance  7 cm
chamber  normal chamber without chamber saturation
sample  0.3 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 270 nm
Description
Featured Industry Chemicals and Industrial Polymers
suitability application for TLC