TLC Analysis of Pyrimidine Derivatives on HPTLC NH2 F254s (analyte set 2)
TLC Analysis of Pyrimidine Derivatives on HPTLC NH2 F254s (analyte set 2)
Conditions
plate HPTLC pre-coated plate NH2 F 254s (1.13192.0001, 1.15647.0001)
eluent [A] chloroform; [B] methanol; (50:50, A:B) 80% saturated with water
migration distance 7 cm
chamber normal chamber without chamber saturation
sample 0.3 μL
detector in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Life Science and Biopharma
suitability application for TLC