TLC Analysis of Pyrimidine Derivatives on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 1)
TLC Analysis of Pyrimidine Derivatives on HPTLC Silica Gel 60 F 254 with Concentrating Zone (analyte set 1)
Conditions
plate  TLC pre-coated plate silica gel 60 F 254 with concentrating zone (1.13728.0001, 1.13727.0001)
eluent [A] chloroform; [B] methanol; (90:10, A:B)
migration distance  5 cm
chamber  normal chamber without chamber saturation
sample  2 μL
detector  in situ evaluation with TLC/HPTLC scanner (Camag) UV 265 nm