TLC Analysis of Substituted Benzoic Acids by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-8 F254s (analyte set 1)
TLC Analysis of Substituted Benzoic Acids by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-8 F254s (analyte set 1)
Conditions
plate TLC pre-coated plate RP-8 F 254s (1.15388.0001, 1.15424.0001)
eluent [A] acetone; [B] water; (40:60, A:B)
(a. Without IPR b. With addition of 0,01 M tetrapropylammonium bromide )
migration distance 7 cm
chamber normal chamber without chamber saturation
detector dipped in dichlorofluorescein (0.2% in ethanol)
(in situ evaluation with TLC/HPTLC scanner (Camag) UV 254 nm)
Description
Featured Industry Chemicals and Industrial Polymers
suitability application for TLC