TLC Analysis of Substituted Benzoic Acids by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-8 F254s (analyte set 3)
TLC Analysis of Substituted Benzoic Acids by Ion-Pair Chromatography on HPTLC Silica Gel 60 RP-8 F254s (analyte set 3)
Conditions
plate TLC pre-coated plate RP-8 F 254s (1.15388.0001, 1.15424.0001)
eluent [A] acetone; [B] water; (40:60, A:B) (a. Without IPR b. With addition of 0,01 M
ammonium bromide c. With addition of 0,01 M tetrabutylammonium bromide)
migration distance 7 cm
chamber  normal chamber without chamber saturation
detector in situ-evaluation with TLC/HPTLC scanner (Camag) UV 254 nm
Description
Featured Industry Chemicals and Industrial Polymers
suitability application for TLC